missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Global/ICH Precious Metals 1mp B (without Os), SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC ICHTXM4
Comprend: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Ir | 100mg/kg |
| Pd | 100mg/kg |
| Pt | 100mg/kg |
| Rh | 100mg/kg |
| Ru | 100mg/kg |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Spécifications
| Precious Metals Imp B (without Os) | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| 15% HCl | |
| 100mg/kg |
| Triple-leached LDPE bottle | |
| AA, ICP | |
| Certified Reference Material | |
| 125 mL |
Product intended for laboratory use only