Learn More
Global/ICH Oral Elemental Impurities A, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier:  SPEX CertiPrep Group LLC ICHTXM2
Numéro de catalogue. ICHTXM2
Il en reste null
Découvrez plus d'offres spéciales
Comprend: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
AnalyteConcentration
Cd25mg/kg
Hg15mg/kg
Pb5mg/kg
As1.5mg/kg
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Spécifications
Spécifications
Oral Elemental Impurities A
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
5% HNO3
Various
Triple-leached LDPE bottle
AA, ICP
Certified Reference Material
125 mL
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS
Documents
Documents
Promotions
Promotions
Product intended for laboratory use only