missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gilson™ PIPETMANTM L Micro-volume Kit
PIPETMANTM L Micro-volume Kit is designed for molecular biology applications.
Supplier: Gilson™ F167550
Description
The PIPETMAN L Micro-volume Kit offers the PIPETMAN P2L and P10L, ideal for pipetting enzymes and reagents; and a PIPETMAN P100L, suited for small-volume dispensing of aqueous fluids of moderate viscosity and density (molecular biology, immunology, etc.).
Kit Contents:
- 3 PIPETMAN L Pipettes: P2L, P10L, and P100L with Gilson certificates
- 2 racks of certified-quality PIPETMAN DIAMOND Tips: DL10 and D200
- 3 SINGLETM pipette holders
- 3 Plastic tip ejectors
- 1 Gilson guide to pipetting
- 1 Two-minute inspection poster
Spécifications
| DL10 and D200 Tips | |
| Blue/White | |
| Manual Air Displacement Pipette | |
| Polypropylene | |
| Pipetman | |
| Starter Kit, Micro | |
| Manual Air Displacement Pipette | |
| 3 | |
| 14.57 in. | |
| 12.2 in. | |
| 4.72 in. | |
| Certified Quality |
| Autoclavable | |
| Reusable | |
| Ergonomic | |
| 1 | |
| Non-sterile | |
| 0.2 to 100 μL | |
| P2L / P10L / P100L, D10 TIPACK, D200 TIPACK | |
| 1 Year | |
| 37 cm | |
| 31 cm | |
| 12 cm |