missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human TGF-beta 3 Recombinant Protein
Recombinant Protein
Supplier: Gibco™ PHG9305
Description
Purity: >98% by SDS-PAGE and HPLC, Endotoxin Concentration: <0.1 ng/μg, Protein Length: 112 aa (2 identical subunits), Biological Activity: ED50 ≤0.05 ng/mL, Molecular Weight: 17.5 kDa
Transforming growth factor beta 3 (TGF beta 3) is the third member of the transforming growth factor family of cytokines, which also includes TGF beta 1 and beta 2. These cytokines are secreted in precursor form consisting of a bioactive C-terminal domain attached to an N-terminal domain known as latency associated protein (LAP). Cleavage of LAP results in the mature protein, which functions as a disulfide-linked homodimer. As with all members of the family, TGF beta 3 is highly conserved across species, with mouse and human TGF beta 3 demonstrating 100% sequence homology and cross-species activity.The members of this family can be expressed by most cell types and exert pleiotropic effects, which include the suppression of B- and T-cell effector activity, mediation of tissue healing, and suppression of tumor proliferation. The promotion of CD4+CD25+ T-cell expansion is a newly discovered function of the TGF beta cytokines, and indicates an important role in the protection against autoimmunity.Spécifications
| P10600 | |
| 0.12% acetic acid with 20% ethanol and no preservative | |
| 25.4 kDa | |
| Sequential Chromatography | |
| Human | |
| ARVD; ARVD1; FLJ16571; H-TGF-b-3; LAP; Latency-associated peptide; LDS5; prepro-transforming growth factor beta-3; RNHF; TGF beta 3; Tgfb3; TGF-B3; Tgfb-3; TGF-beta3; TGF-beta-3; Transforming growth factor; transforming growth factor beta 3; transforming growth factor beta precursor; transforming growth factor beta-3; Transforming growth factor beta-3 proprotein; transforming growth factor, beta 3; transforming growth factor-beta3 | |
| ED50 ≤ 0.05 ng/mL (≥ 12.5 x 10e6 IU/mg); determined by a toxicity assay using the WHO Standard 98/608 for direct comparison. | |
| Recombinant | |
| 4°C | |
| Growth Factors & Receptors | |
| TGFs (Transforming Growth Factors) |
| Control, ELISA, Functional Assay, Immunohistochemistry, Western Blot | |
| 7043 | |
| Human TGF-beta 3 | |
| 5 μg | |
| <0.1 ng/ μg | |
| TGF beta-3 | |
| Unconjugated | |
| Human TGF-beta 3 recombinant protein is a homodimer containing two 112 amino acid subunits | |
| E. coli | |
| Liquid | |
| >98% by SDS-PAGE and HPLC |