missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ DMEM, high glucose, HEPES DFS Item

DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells.

Supplier:  Gibco™ 12430047

 View more versions of this product

Catalog No. 12430047


Only null left
Explore available promotions

Description

Description

DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This DMEM is modified as follows:

With: High Glucose, L-glutamine, HEPES, Phenol Red

Without: Sodium Pyruvate

Using DMEM

DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate. DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Specifications

Specifications

HeLa, 293, Cos-7, and PC-12
Animal Origin-free
Liquid
Standard Serum Supplementation
Sterile-filtered
No Sodium Pyruvate
cGMP-compliant under the ISO 13485 standard
1000 mL
Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Primary Fibroblasts, Neurons, Glial Cells, HUVECs, Smooth Muscle Cells
1 X
DMEM (Dulbecco's Modified Eagle Medium)
Sterile-filtered
High Glucose, Glutamine, HEPES, Phenol Red
Mammalian cell culture
Gibco
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.