missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva GammaBind™ Plus Sepharose™ Medium
Engineered for better binding to rat and mouse IgG subclasses
$39180.62 - $39180.62
Spécifications
| Product Type | GammaBind Plus Sepharose |
|---|---|
| Ligand | Recombinant GammaBind G type 3 lacking albumin-binding region |
| Particle Size | 90 μm |
| Content And Storage | 2°C to 8°C, 20% Ethanol |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
45-500-106
|
Cytiva
17088604 |
500 mL |
N/A
|
N/A | |||||
| Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
Description
- Recombinant Protein G type 3 coupled to Sepharose CL-6B for easy binding and fast separation
- More enhanced binding capabilities than GammaBind G at both laboratory and process scale
- Yields up to 18mg/mL mouse IgG
- Binds to all IgG subclasses from human, mouse, and rat; binds total IgG from guinea pig, rabbit, goat, cow, sheep, and horse
Spécifications
| GammaBind Plus Sepharose | |
| Recombinant GammaBind G type 3 lacking albumin-binding region | |
| 90 μm | |
| 2°C to 8°C, 20% Ethanol |