Learn More
(Formylmethylene)triphenylphosphorane, 97%, may cont. up to ca 3% water
CAS: 2136-75-6 | C20H17OP | 304.329 g/mol
Supplier: Thermo Scientific Chemicals A1170914
Description
(Formylmethylene)triphenylphosphorane is used as Wittig reagent. It is also used as Pharmaceutical Intermediates. It is employed as raw material in organic Synthesis, Agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(Formylmethylene)triphenylphosphorane is used as Wittig reagent. It is also used as Pharmaceutical Intermediates. It is employed as raw material in organic Synthesis, Agrochemicals.
Solubility
Soluble in Chloroform, Methanol.
Notes
Air Sensitive. Store at -20°C. Incompatible with strong oxidizing agents.
Spécifications
| (Formylmethylene)triphenylphosphorane | |
| 183°C to 188°C | |
| MFCD00006994 | |
| 523797 | |
| 2-triphenylphosphoranylidene acetaldehyde, formylmethylene triphenylphosphorane, triphenylphosphoranylidene acetaldehyde, formylmethylenetriphenylphosphorane, acetaldehyde, triphenylphosphoranylidene, 2-triphenyl-??-phosphanylidene acetaldehyde, acmc-209fjg, ksc492c5l | |
| CQCAYWAIRTVXIY-UHFFFAOYSA-N | |
| 2-(triphenyl-$l^{5}-phosphanylidene)acetaldehyde | |
| 75051 | |
| 97% |
| 2136-75-6 | |
| C20H17OP | |
| 25 g | |
| Air sensitive | |
| Soluble in Chloroform,Methanol. | |
| C1=CC=C(C=C1)P(=CC=O)(C2=CC=CC=C2)C3=CC=CC=C3 | |
| 304.329 | |
| 304.33 | |
| May contain up to ≈3% water |
Identifiants chimiques
| 2136-75-6 | |
| 304.329 | |
| CQCAYWAIRTVXIY-UHFFFAOYSA-N | |
| 75051 | |
| C1=CC=C(C=C1)P(=CC=O)(C2=CC=CC=C2)C3=CC=CC=C3 |
| C20H17OP | |
| MFCD00006994 | |
| 2-triphenylphosphoranylidene acetaldehyde, formylmethylene triphenylphosphorane, triphenylphosphoranylidene acetaldehyde, formylmethylenetriphenylphosphorane, acetaldehyde, triphenylphosphoranylidene, 2-triphenyl-??-phosphanylidene acetaldehyde, acmc-209fjg, ksc492c5l | |
| 2-(triphenyl-$l^{5}-phosphanylidene)acetaldehyde |
Sécurité et manipulation
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 218-375-6
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only