missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde Solution, 8% v/v, Reagents

Formaldehyde Solution, 8% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Labchem Inc CF1006004A

 Afficher plus de versions de ce produit

Numéro de catalogue. 01-335-277


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Spécifications

Spécifications

Formaldehyde Solution, 8% v/v
Colorless
Poly Bottle
4 L
Formalin
Reagent
7732-18-5,50-00-0
100°C
HCHO/H2O
1.01
8%
Liquid
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions