missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde Solution, 8% v/v, Reagents

Formaldehyde Solution, 8% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

$323.54 - $562.75

Specifications

Chemical Name or Material Formaldehyde Solution, 8% v/v
CAS 7732-18-5,50-00-0
Color Colorless
Boiling Point 100°C
Packaging Poly Bottle
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01-335-278
View Documents
Labchem Inc
CF1006001A
1 L
Each for $323.54
Only null left
 
01-335-277
View Documents
Labchem Inc
CF1006004A
4 L
Each for $562.75
Only null left
 
Description

Description

Specifications

Specifications

Formaldehyde Solution, 8% v/v
Colorless
Poly Bottle
1.01
8%
Liquid
7732-18-5,50-00-0
100°C
HCHO/H2O
Formalin
Reagent
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'