missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Folin-Ciocalteu Phenol TS Test Solution, USP, Spectrum™ Chemical
F-184
$368.71 - $1417.04
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | H4Na2O6W |
| Molecular Weight (g/mol) | 329.85 |
| InChI Key | WPZFLQRLSGVIAA-UHFFFAOYSA-N |
| IUPAC Name | disodium dioxotungstenbis(olate) dihydrate |
| SMILES | O.O.[Na+].[Na+].[O-][W]([O-])(=O)=O |
Description
Spectrum™ Chemical Folin-Ciocalteu Phenol TS, USP Test Solution is a solution, sometimes designated with 'TS or Test Solution', that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.
Chemical Identifiers
| 7732-18-5 | |
| 329.85 | |
| disodium dioxotungstenbis(olate) dihydrate |
| H4Na2O6W | |
| WPZFLQRLSGVIAA-UHFFFAOYSA-N | |
| O.O.[Na+].[Na+].[O-][W]([O-])(=O)=O |
Specifications
| 7732-18-5 , 10377-48-7 , 7664-38-2 , 7647-01-0 , 10213-10-2 , 10102-40-6 | |
| H4Na2O6W | |
| WPZFLQRLSGVIAA-UHFFFAOYSA-N | |
| disodium dioxotungstenbis(olate) dihydrate | |
| Chemical Solution | |
| Acidic | |
| Folin-Ciocalteu Phenol TS |
| 80%,<10%,<5%,<5%,<3%,<3% | |
| UN3264 | |
| O.O.[Na+].[Na+].[O-][W]([O-])(=O)=O | |
| 329.85 | |
| Amber Glass Bottle | |
| 100 mL |