missing translation for 'onlineSavingsMsg'
Learn More

Folin-Ciocalteu Phenol TS Test Solution, USP, Spectrum™ Chemical

Catalog No. 18602808
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602808 100 mL Amber Glass Bottle
18602809 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18602808 Supplier Spectrum Chemical Mfg Cor Supplier No. F184100ML
Only null left

F-184

Spectrum™ Chemical Folin-Ciocalteu Phenol TS, USP Test Solution is a solution, sometimes designated with 'TS or Test Solution', that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.

Chemical Identifiers

CAS 7732-18-5 , 10377-48-7 , 7664-38-2 , 7647-01-0 , 10213-10-2 , 10102-40-6
Molecular Formula H4Na2O6W
Molecular Weight (g/mol) 329.85
InChI Key WPZFLQRLSGVIAA-UHFFFAOYSA-N
IUPAC Name disodium dioxotungstenbis(olate) dihydrate
SMILES O.O.[Na+].[Na+].[O-][W]([O-])(=O)=O

Specifications

CAS Max % 80%, <10%, <5%, <5%, <3%, <3%
UN Number UN3264
Grade Chemical Solution
Packaging Amber Glass Bottle
pH Acidic
Quantity 100 mL
Chemical Name or Material Folin-Ciocalteu Phenol TS