accessibility menu, dialog, popup

UserName

H2OH2ONaNaOWOOO

CAS RN 10213-10-2

IUPAC Name: disodium dioxotungstenbis(olate) dihydrate
Synonyms: sodium tungstate dihydrate na2WO4.2H2O sodium tungstate(VI) dihydrate sodium orthotungstate dihydrate tungstic acid, sodium salt, dihydrate
Molecular Weight (g/mol): 329.85
Molecular Formula: H4Na2O6W
InChi Key: WPZFLQRLSGVIAA-UHFFFAOYSA-N
SMILES: O.O.[Na+].[Na+].[O-][W]([O-])(=O)=O
Grade 
  • (2)
  • (3)
  • (4)
  • (6)
  • (2)
Molecular Weight (g/mol) 
  • (6)
  • (7)
  • (4)
Percent Purity 
  • (9)
  • (4)
  • (2)
  • (2)
Physical Form 
  • (3)
  • (4)
  • (1)
  • (4)
  • (2)
  • (3)
Quantity 
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (4)
  • (2)
  • (4)
  • (11)
  • (2)
  • (3)
  • (4)
  • (6)
  • (2)
  • (6)
  • (7)
  • (4)
  • (9)
  • (4)
  • (2)
  • (2)
  • (3)
  • (4)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (5)