accessibility menu, dialog, popup

UserName

LiLiOSOOO

CAS RN 10377-48-7

IUPAC Name: dilithium(1+) sulfate
Synonyms: lithium sulfate|lithium sulphate|Li2SO4|lithiophor|dilthium sulfate
Molecular Weight (g/mol): 109.94
Molecular Formula: Li2O4S
InChi Key: INHCSSUBVCNVSK-UHFFFAOYSA-L
SMILES: [Li+].[Li+].[O-]S([O-])(=O)=O
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
Grade 
  • (2)
  • (3)
  • (2)
  • (4)
Percent Purity 
  • (4)
  • (3)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (11)
  • (4)
  • (12)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (2)
  • (3)
  • (2)
  • (4)
  • (4)
  • (3)
  • (2)
  • (2)