missing translation for 'onlineSavingsMsg'
Learn More

Fluoride Standard, 10ppm F- Premixed with Equal Volume of TISAB II, Certified, 10.0ppm ±0.1ppm, LabChem™

$114.79 - $505.25

Chemical Identifiers

CAS 7681-49-4
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
IUPAC Name sodium fluoride
SMILES [F-].[Na+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
LC146401
View Documents
LabChem
LC146401
500 mL Poly Bottle
Each for $114.79
Only null left
 
LC146402
View Documents
LabChem
LC146402
1 L Poly Bottle
Each for $203.57
Only null left
 
LC146404
View Documents
LabChem
LC146404
4 L Poly Bottle
Each for $505.25
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7681-49-4
41.99
PUZPDOWCWNUUKD-UHFFFAOYSA-M
[F-].[Na+]
FNa
MFCD00003524
sodium fluoride

Specifications

Poly Bottle
91.839,2.9,2.85,2.2,0.2
Liquid
10.0 ppm ±0.1 ppm
MFCD00003524
Soluble in water
[F-].[Na+]
41.99
41.99
Fluoride Standard, 10ppm F- Premixed with Equal Volume of TISAB II
7681-49-4 , 7647-14-5 , 64-19-7 , 1310-73-2 , 125572-95-4 , 7732-18-5
Passes Test
FNa
NaF
Colorless
PUZPDOWCWNUUKD-UHFFFAOYSA-M
sodium fluoride
500 mL
Certified
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents