accessibility menu, dialog, popup

UserName

CAS RN 125572-95-4

IUPAC Name: 2-{[(1R,2R)-2-[bis(carboxylatomethyl)azaniumyl]cyclohexyl](carboxylatomethyl)azaniumyl}acetate
Synonyms: 2-{[(1R,2R)-2-[bis(carboxylatomethyl)ammonio]cyclohexyl](carboxylatomethyl)ammonio}acetate
Molecular Weight (g/mol): 344.32
Molecular Formula: C14H20N2O8
InChi Key: FCKYPQBAHLOOJQ-NXEZZACHSA-L
SMILES: [O-]C(=O)C[NH+](CC([O-])=O)[C@@H]1CCCC[C@H]1[NH+](CC([O-])=O)CC([O-])=O
Quantity 
  • (5)
  • (1)
  • (1)
  • (6)
  • (1)
  • (5)
Percent Purity 
  • (3)
  • (4)
  • (9)
Grade 
  • (7)
  • (2)
  • (4)
Physical Form 
  • (3)
  • (3)
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (4)
  • (3)
  • (4)
  • (3)
  • (9)
  • (5)
  • (1)
  • (1)
  • (6)
  • (1)
  • (5)
  • (3)
  • (4)
  • (9)
  • (7)
  • (2)
  • (4)
  • (3)
  • (3)
  • (7)