Learn More
BUCHI FlashPure EcoFlex Flash Chromatography Cartridges: C18
FlashPure EcoFlex cartridges are designed to optimized backpressure, price and performance.
Supplier:  BUCHI 140000056
 Afficher plus de versions de ce produit
Numéro de catalogue. 05410030
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Most economical solution without sacrificing quality for standard separations
  • Large selection of cartridge sizes from 4g-5,000g
  • Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen
  • Silica is ideal for basic applications and polar compounds using organic solvents
  • Amino functionalized silica provides better peak shapes with polar and basic compounds and is ideal for carbohydrates
  • C18 functionalized silica is ideal for peptides and non-polar compounds in an aqueous mobile phase
  • Diol functionalized silica provides an alternate selectivity for a variety of polarity types and is ideal for lipids
  • Neutral Alumina is ideal for aldehydes, ketones, and esters
Spécifications
Spécifications
Polypropylene
1 Pc.
800 g
Non-Polar Compounds, Ideal for Peptides
C18
40 to 60 μm
C18
Any Flash System
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS