missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BUCHI FlashPure EcoFlex Flash Chromatography Cartridges: C18
Description
- Most economical solution without sacrificing quality for standard separations
- Large selection of cartridge sizes from 4g-5,000g
- Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen
- Silica is ideal for basic applications and polar compounds using organic solvents
- Amino functionalized silica provides better peak shapes with polar and basic compounds and is ideal for carbohydrates
- C18 functionalized silica is ideal for peptides and non-polar compounds in an aqueous mobile phase
- Diol functionalized silica provides an alternate selectivity for a variety of polarity types and is ideal for lipids
- Neutral Alumina is ideal for aldehydes, ketones, and esters
Specifications
Specifications
| Material | Polypropylene |
| Type | C18 |
| Includes | 2 Pc. |
| Resolution | 40 to 60 μm |
| Bed Weight (Metric) | 120 g |
| Column Type | C18 |
| For Use With (Application) | Non-Polar Compounds, Ideal for Peptides |
| For Use With (Equipment) | Any Flash System |