Learn More
BUCHI FlashPure Chromatography Solid Loader
Numéro de catalogue. 05405537
Change view
missing translation for 'orderingAttributeHoverText'
Size (Metric) Tube Rack:
12 g
120 g
15 g
20 g
40 g
80 g
Numéro de catalogue. Size (Metric) Tube Rack
05405537 15 g
05402905 12 g
05402906 20 g
05402907 40 g
05410123 80 g
05410124 120 g
6 options
Numéro de catalogue. 05405537
Supplier: BUCHI 11068975
Il en reste null
FlashPure Empty Solid Loaders are ideal for samples that are not readily soluble in the desired mobile phase when performing flash chromatography. The sample is adsorbed onto a solid phase (silica or Celite) and then added to the empty solid loader tube.
  • Screw cap makes sealing easy
  • Various sizes for differing sample amounts
  • Choose a size large enough for the sample
  • Insertion Rod allows frit placement and loading of samples smaller than the entire volume
Spécifications
Type Chromatography Loader
Includes Adapter Set, Sleeve, 20 Tubes, and 40 Frits
Size (Metric) Tube Rack 15 g
For Use With (Equipment) Pure Chromatography Systems