missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP3 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$344.88 - $344.88
Spécifications
| Antigen | FKBP3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
501562997
|
Bethyl Laboratories Inc
A302601AT |
20 μL |
chaque for $344.88
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Bovine and Rabbit
The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. This encoded protein is a cis-trans prolyl isomerase that binds the immunosuppressants FK506 and rapamycin, as well as histone deacetylases, the transcription factor YY1, casein kinase II, and nucleolin. It has a higher affinity for rapamycin than for FK506 and thus may be an important target molecule for immunosuppression by rapamycin.Spécifications
| FKBP3 | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| FKBP-25, PPIase, 25 kDa FK506-binding protein; 25 kDa FKBP; FK506-binding protein 25, T-cell; FK506-binding protein 3 (25kD); FKBP-3; PPIase FKBP3; immunophilin FKBP25; peptidyl-prolyl cis-trans isomerase FKBP3; rapamycin binding protein; rapamycin-selective 25 kDa immunophilin; rotamase | |
| FKBP3 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q00688 | |
| 2287 | |
| Between 100 and 150 | |
| Primary | |
| 4° C | |
| FKBP3 |