Learn More
Thermo Scientific Chemicals Fipronil
CAS: 120068-37-3 | C12H4Cl2F6N4OS | 437.14 g/mol
$367.25 - $367.25
Identifiants chimiques
| CAS | 120068-37-3 |
|---|---|
| Molecular Formula | C12H4Cl2F6N4OS |
| Molecular Weight (g/mol) | 437.14 |
| InChI Key | ZOCSXAVNDGMNBV-UHFFFAOYNA-N |
| IUPAC Name | 5-amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-trifluoromethanesulfinyl-1H-pyrazole-3-carbonitrile |
| SMILES | NC1=C(C(=NN1C1=C(Cl)C=C(C=C1Cl)C(F)(F)F)C#N)S(=O)C(F)(F)F |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
AC465570250
|
Thermo Scientific Chemicals
465570250 |
25 g |
chaque for $367.25
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 120068-37-3 | |
| 437.14 | |
| 5-amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-trifluoromethanesulfinyl-1H-pyrazole-3-carbonitrile |
| C12H4Cl2F6N4OS | |
| ZOCSXAVNDGMNBV-UHFFFAOYNA-N | |
| NC1=C(C(=NN1C1=C(Cl)C=C(C=C1Cl)C(F)(F)F)C#N)S(=O)C(F)(F)F |
Spécifications
| 120068-37-3 | |
| C12H4Cl2F6N4OS | |
| Solubility aq. soln.: insoluble. Other solubilities: soluble in acetone, acetonitrile, insoluble in toluene, hexane | |
| NC1=C(C(=NN1C1=C(Cl)C=C(C=C1Cl)C(F)(F)F)C#N)S(=O)C(F)(F)F | |
| 437.14 | |
| Fipronil |
| 200.0°C to 201.0°C | |
| 25 g | |
| ZOCSXAVNDGMNBV-UHFFFAOYNA-N | |
| 5-amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-trifluoromethanesulfinyl-1H-pyrazole-3-carbonitrile | |
| 437.15 |
Sécurité et manipulation
GHS H Statement Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
RUO – Research Use Only