missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fipronil

CAS: 120068-37-3 | C12H4Cl2F6N4OS | 437.14 g/mol

Supplier:  Thermo Scientific Chemicals 465570250

 Afficher plus de versions de ce produit

Numéro de catalogue. AC465570250


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

Fipronil
200.0°C to 201.0°C
25 g
ZOCSXAVNDGMNBV-UHFFFAOYNA-N
5-amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-trifluoromethanesulfinyl-1H-pyrazole-3-carbonitrile
437.15
120068-37-3
C12H4Cl2F6N4OS
Solubility aq. soln.: insoluble. Other solubilities: soluble in acetone, acetonitrile, insoluble in toluene, hexane
NC1=C(C(=NN1C1=C(Cl)C=C(C=C1Cl)C(F)(F)F)C#N)S(=O)C(F)(F)F
437.14

Identifiants chimiques

120068-37-3
437.14
5-amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-trifluoromethanesulfinyl-1H-pyrazole-3-carbonitrile
C12H4Cl2F6N4OS
ZOCSXAVNDGMNBV-UHFFFAOYNA-N
NC1=C(C(=NN1C1=C(Cl)C=C(C=C1Cl)C(F)(F)F)C#N)S(=O)C(F)(F)F
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement Toxic if swallowed.

Toxic in contact with skin.

Toxic if inhaled.

Causes damage to organs through prolonged or repeated exposure.

Very toxic to aquatic life with long lasting effects.

GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

Wear protective gloves/protective clothing.

IF ON SKIN: Wash with plenty of soap and water.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only