Learn More
Thermo Scientific™ BseNI (BsrI) (10 U/μL)
Cut at ACTGG(1/-1)^ sites with BseNI (BsrI) restriction enzyme, which performs best at 65°C in B buffer (Isoschizomers: Bse1I, BsrI, BsrSI).
$145.28 - $145.28
Spécifications
Concentration 10 U/μL
voir les spécifications
Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
FERER0881
Afficher les documents
Thermo Scientific™
ER0881
1,000 units
chaque for $145.28
Il en reste null
Description
Description
Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
Highlights
  • Superior quality -stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
Spécifications
Spécifications
10 U/μL
Videos
FDS
missing translation for 'documents'
missing translation for 'documents'
For Research Use Only. Not for use in diagnostic procedures.