Learn More
Falcon™ HTS 24-Well Multiwell Permeable Support System
Save time and labor in HTS protocols—grow, handle, and analyze 24 samples simultaneously. Falcon™ HTS Multiwell Insert System features a generous, automation-compatible sampling port which allows sampling above and below membranes.
$1777.70 - $2014.87
Spécifications
| Color | Clear |
|---|---|
| Culture Area | 0.3 cm2 |
| Sterility | Sterile |
| Type | Multi-well Insert System |
| Well Count | 24 |
Comprend: 24-well insert plate, 24-well plate or single-well feeder tray, and lid
| Numéro de catalogue | Numéro du manufacturier. | Pore Size | Quantity | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Pore Size | Quantity | Prix | Quantité | ||||
|
08-780
|
Falcon™
351185 |
8 μm | 5/Cs. |
caisse de 5 chaque for $1,873.62
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
|
08-778
|
Falcon™
351183 |
3 μm | 5/Cs. |
caisse de 5 chaque for $2,014.87
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
|
08-776
|
Falcon™
351181 |
1 μm | 5/Cs. |
caisse de 5 chaque for $1,777.70
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
Falcon 24 Multiwell Insert Systems are designed to automate many commonly used cell-based assays for drug discovery. Available in a choice of membrane pore sizes, 24 Multiwell inserts have been successfully used for a variety of applications including permeability studies for oral bioavailability (e.g., Caco-2 cells), chemotaxis, cell migration and invasion assays. These insert systems offer all the benefits of Falcon Individual Cell Culture Inserts in an automation-friendly format that is compatible with most robots and fluid handlers. The Falcon 24 Multiwell Insert Plate is a single unit that is compatible with all Falcon 24 well plates and Feeder Tray.
Spécifications
| Clear | |
| Sterile | |
| 24 | |
| Polyethylene Terephthalate |
| 0.3 cm2 | |
| Multi-well Insert System | |
| Excellent for differentiation, toxicology, migration and invasion studies |