missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EXPRESS qPCR Supermix, with premixed ROX
EXPRESS qPCR SuperMix kits are compatible with fast-cycling instruments for your probe-based qPCR assays.
$1,754.00 - $6,840.00
Spécifications
| Content And Storage | EXPRESS qPCR SuperMix kits are provided as a 2X qPCR SuperMix with ROX premixed or in a separate tube. Store at -20°C upon receipt. Once thawed, the EXPRESS qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteedstable for 6 months when properly stored. Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
|---|---|
| Detection Method | Primer-probe |
| Format | Tube |
| GC-Rich PCR Performance | High |
| PCR Method | qPCR |
| Numéro de catalogue | Numéro du manufacturier. | No. of Reactions | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | No. of Reactions | Prix | Quantity & Availability | |||||
11795200
|
Invitrogen™
11795200 |
200 Reactions |
chaque for $1,754.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
1179501K
|
Invitrogen™
1179501K |
1000 Reactions |
chaque for $6,840.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
- Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
- UDG carryover protection - uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination
Order Info
Guaranteed minimum shelf life is 60 days.
Spécifications
| EXPRESS qPCR SuperMix kits are provided as a 2X qPCR SuperMix with ROX premixed or in a separate tube. Store at -20°C upon receipt. Once thawed, the EXPRESS qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteedstable for 6 months when properly stored. Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
|
| Tube | |
| qPCR | |
| 7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System, Eppendorf™ MasterCycler™ RealPlex, StepOne™, Fast Mode, StepOne™, Standard Mode | |
| Gene Expression |
| Primer-probe | |
| High | |
| Fast or Standard | |
| Express qPCR Supermix | |
| DNA (Genomic), cDNA |
For Research Use Only. Not for use in diagnostic procedures.