missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EXPRESS qPCR Supermix, with premixed ROX
EXPRESS qPCR SuperMix kits are compatible with fast-cycling instruments for your probe-based qPCR assays.
Supplier: Invitrogen™ 1179501K
Description
- Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
- UDG carryover protection - uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination
Order Info
Guaranteed minimum shelf life is 60 days.
Chemical Identifiers
| EXPRESS qPCR SuperMix kits are provided as a 2X qPCR SuperMix with ROX premixed or in a separate tube. Store at -20°C upon receipt. Once thawed, the EXPRESS qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteedstable for 6 months when properly stored. Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
|
| Tube | |
| qPCR | |
| 7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System, Eppendorf™ MasterCycler™ RealPlex, StepOne™, Fast Mode, StepOne™, Standard Mode | |
| Gene Expression |
| Primer-probe | |
| High | |
| Fast or Standard | |
| Express qPCR Supermix | |
| DNA (Genomic), cDNA |
Specifications
| EXPRESS qPCR SuperMix kits are provided as a 2X qPCR SuperMix with ROX premixed or in a separate tube. Store at -20°C upon receipt. Once thawed, the EXPRESS qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteedstable for 6 months when properly stored. Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
|
| Tube | |
| qPCR | |
| Fast or Standard | |
| Express qPCR Supermix | |
| 1000 Reactions | |
| 1000 Reactions |
| Primer-probe | |
| High | |
| Taq DNA Polymerase | |
| 7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System, Eppendorf™ MasterCycler™ RealPlex, StepOne™, Fast Mode, StepOne™, Standard Mode | |
| 1,000 reactions | |
| Gene Expression | |
| DNA (Genomic), cDNA |
For Research Use Only. Not for use in diagnostic procedures.