missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Evodiamine

CAS: 518-17-2 | C19H17N3O, C19H17N3O | 303.37 g/mol

Supplier:  Thermo Scientific Chemicals J62771MC

Quantity 100 mg

 Afficher plus de versions de ce produit

Numéro de catalogue. AAJ62771MC


Il en reste null

Description

Description

A vanilloid receptor agonist that induces apoptosis in leukemia U937 cells

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

518-17-2
C19H17N3O

Spécifications

100 mg
-30°C to -10°C
518-17-2
303.40
Solid
CN1[C@H]2N(CCC3=C2NC2=CC=CC=C32)C(=O)C2=CC=CC=C12
303.37
Off-white to pale yellow
A vanilloid receptor agonist that induces apoptosis in leukemia U937 cells
C19H17N3O
Evodiamine
TXDUTHBFYKGSAH-SFHVURJKSA-N
(1S)-21-methyl-3,13,21-triazapentacyclo[11.8.0.0²,¹⁰.0⁴,⁹.0¹⁵,²⁰]henicosa-2(10),4,6,8,15,17,19-heptaen-14-one
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

Recommended Storage : Store at -20°C

FDS
Documents

Documents

Promotions

Promotions