missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Evodiamine
CAS: 518-17-2 | C19H17N3O, C19H17N3O | 303.37 g/mol
$121.52 - $219.36
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAJ62771MD
|
Thermo Scientific Chemicals
J62771MD |
250 mg |
Each for $219.36
|
|
|||||
|
AAJ62771MC
|
Thermo Scientific Chemicals
J62771MC |
100 mg |
Each for $121.52
|
|
|||||
Description
A vanilloid receptor agonist that induces apoptosis in leukemia U937 cells
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 518-17-2 | |
| C19H17N3O |
Specifications
| Off-white to pale yellow | |
| A vanilloid receptor agonist that induces apoptosis in leukemia U937 cells | |
| C19H17N3O | |
| Evodiamine | |
| TXDUTHBFYKGSAH-SFHVURJKSA-N | |
| (1S)-21-methyl-3,13,21-triazapentacyclo[11.8.0.0²,¹⁰.0⁴,⁹.0¹⁵,²⁰]henicosa-2(10),4,6,8,15,17,19-heptaen-14-one |
| -30°C to -10°C | |
| 518-17-2 | |
| 303.40 | |
| Solid | |
| CN1[C@H]2N(CCC3=C2NC2=CC=CC=C32)C(=O)C2=CC=CC=C12 | |
| 303.37 |
Safety and Handling
Recommended Storage : Store at -20°C
RUO – Research Use Only