Learn More
Cytiva ETFE Ferrule
Ferrules used for chromatography tubing. Cytiva Lifescience™ ETFE Ferrule is made from ethylenetetra fluoroethylene and comes with different outside diameters.
Supplier: Cytiva 18112118
Description
- Ferrules
- Chromatography accessory
- Used for: protecting the end of tubing
- Packsize: 10 Connections
- Ethylenetetra fluoroethylene (ETFE)
Spécifications
| Tubing | |
| ETFE | |
| 10 Pack |
| 1/8 in. | |
| Ferrule |