missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ ES 7X™-PF Cleaning Solution
GREENER_CHOICE

ES 7X™-PF Cleaning Solution is a Phosphate Free detergent, specifically developed for safe and effective cleaning of laboratory instruments and glassware.

$211.00 - $6,328.96


Produits 4
Numéro de catalogue Numéro du manufacturier. Volume (English) Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Volume (English) Prix Quantity & Availability  
ICN7667194
GREENER_CHOICE
Afficher les documents Détails de la promotion
MP Biomedicals™
097667194
4 x 1 gal.
chaque for $480.40
Il en reste null
 
ICN7667195
GREENER_CHOICE
Afficher les documents Détails de la promotion
MP Biomedicals™
097667195
5 gal.
chaque for $543.17
Il en reste null
 
ICN7667193
GREENER_CHOICE
Afficher les documents Détails de la promotion
MP Biomedicals™
097667193
1 gal.
chaque for $211.00
Il en reste null
 
ICN7667198
GREENER_CHOICE
Afficher les documents Détails de la promotion
MP Biomedicals™
097667198
55 gal.
chaque for $6,328.96
Il en reste null
 
Description

Description

  • Environmentally safe and phosphate free.
  • Developed to provide safe and effective cleaning of laboratory instruments and glassware including test tubes, petri-dishes, flasks, microscopy slides, pathology equipment and surgical instruments
  • Several convenient package sizes are available
  • Sold in concentrated form so the customer can prepare the appropriate dilution as needed
  • Completely drains from glassware in seconds
  • Eliminates residue leaving no interfering fluorescence
  • pH neutral, and no need for pH adjustment at any concentration
  • No etch to glass or plastic labware at any concentration
Videos
FDS
Documents

Documents