missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ ES 7X™-PF Cleaning Solution

ES 7X™-PF Cleaning Solution is a Phosphate Free detergent, specifically developed for safe and effective cleaning of laboratory instruments and glassware.
Supplier: MP Biomedicals™ 097667198
Description
- Environmentally safe and phosphate free.
- Developed to provide safe and effective cleaning of laboratory instruments and glassware including test tubes, petri-dishes, flasks, microscopy slides, pathology equipment and surgical instruments
- Several convenient package sizes are available
- Sold in concentrated form so the customer can prepare the appropriate dilution as needed
- Completely drains from glassware in seconds
- Eliminates residue leaving no interfering fluorescence
- pH neutral, and no need for pH adjustment at any concentration
- No etch to glass or plastic labware at any concentration
Specifications
| Liquid | |
| Yellow | |
| 15°C to 30°C | |
| 55 gal. |
| 7 to 8 | |
| 7X | |
| Use on Glass, Plastic, benchtop equipment and tubing |