Learn More
Thermo Scientific Chemicals Erythrosin B, pure, certified
CAS: 16423-68-0 | C20H6I4Na2O5 | 879.86 g/mol
$112.20 - $398.91
Identifiants chimiques
| CAS | 16423-68-0 |
|---|---|
| Molecular Formula | C20H6I4Na2O5 |
| Molecular Weight (g/mol) | 879.86 |
| MDL Number | MFCD00144257 |
| InChI Key | IINNWAYUJNWZRM-UHFFFAOYSA-L |
| Synonym | Acid Red 51, C.I. 45430 |
| PubChem CID | 12961638 |
| SMILES | [Na+].[Na+].[O-]C(=O)C1=C(C=CC=C1)C1=C2C=C(I)C(=O)C(I)=C2OC2=C(I)C([O-])=C(I)C=C12 |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|
AC409450250
|
Thermo Scientific Chemicals
409450250 |
25 g | Glass bottle |
chaque for $112.20
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
|
AC409451000
|
Thermo Scientific Chemicals
409451000 |
100 g | Glass bottle |
chaque for $398.91
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorIdentifiants chimiques
| 16423-68-0 | |
| 879.86 | |
| IINNWAYUJNWZRM-UHFFFAOYSA-L | |
| 12961638 |
| C20H6I4Na2O5 | |
| MFCD00144257 | |
| Acid Red 51, C.I. 45430 | |
| [Na+].[Na+].[O-]C(=O)C1=C(C=CC=C1)C1=C2C=C(I)C(=O)C(I)=C2OC2=C(I)C([O-])=C(I)C=C12 |
Spécifications
| 16423-68-0 | |
| 80% min. | |
| MFCD00144257 | |
| 15, 3749 | |
| IINNWAYUJNWZRM-UHFFFAOYSA-L | |
| [Na+].[Na+].[O-]C(=O)C1=C(C=CC=C1)C1=C2C=C(I)C(=O)C(I)=C2OC2=C(I)C([O-])=C(I)C=C12 | |
| 879.86 | |
| 524 to 527nm | |
| Pure | |
| Brown to Red | |
| Fine Powder |
| 1.15 to 1.55 | |
| C20H6I4Na2O5 | |
| 19, II, 258 | |
| Acid Red 51, C.I. 45430 | |
| Authentic | |
| disodium;2',4',5',7'-tetraiodo-3-oxospiro[2-benzofuran-1,9'-xanthene]-3',6'-diolate | |
| 12961638 | |
| 879.85 | |
| Glass bottle | |
| 25 g | |
| Erythrosin B, Pure |
Sécurité et manipulation
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
RUO – Research Use Only