missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Erythrosin B, pure, certified

CAS: 16423-68-0 | C20H6I4Na2O5 | 879.86 g/mol

Supplier:  Thermo Scientific Chemicals 409451000

 View more versions of this product

Catalog No. AC409451000

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

Erythrosin B
Pure
1.15 to 1.55
C20H6I4Na2O5
19, II, 258
Acid Red 51, C.I. 45430
IINNWAYUJNWZRM-UHFFFAOYSA-L
[Na+].[Na+].[O-]C(=O)C1=C(C=CC=C1)C1=C2C=C(I)C(=O)C(I)=C2OC2=C(I)C([O-])=C(I)C=C12
879.86
524 to 527nm
Pure
Brown to Red
Fine Powder
>300.0°C
16423-68-0
80% min.
MFCD00144257
15, 3749
Solubility in water: soluble. Other solubilities: soluble in alcohol
Authentic
disodium 2-(2,4,5,7-tetraiodo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
12961638
879.85
Glass bottle
100 g

Chemical Identifiers

16423-68-0
879.86
IINNWAYUJNWZRM-UHFFFAOYSA-L
12961638
C20H6I4Na2O5
MFCD00144257
Acid Red 51, C.I. 45430
[Na+].[Na+].[O-]C(=O)C1=C(C=CC=C1)C1=C2C=C(I)C(=O)C(I)=C2OC2=C(I)C([O-])=C(I)C=C12
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only