missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Extran™ 300 Detergent

Powerful, liquid concentrate specially formulated to remove stubborn contamination and burnt-on residues

$131.28 - $380.93

Spécifications

Form Liquid
Appearance Off-White
For Use With (Application) Used on equipment and apparatus made of Glass, Plastic, Quartz, Porcelain, Ferrous Metals and Rubber
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Volume (Metric) Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Volume (Metric) Prix Quantité  
MEX09961
Afficher les documents
MilliporeSigma™
EX09961
4 L
chaque for $380.93
Il en reste null
 
MEX09962
Afficher les documents
MilliporeSigma™
EX09962
1 L
chaque for $131.28
Il en reste null
 
Description

Description

Can be used on equipment and apparatus made of glass, plastic, quartz, porcelain, ferrous metals and rubber.

  • Especially effective at removing organic materials such as blood and proteins
  • Works well in ultrasonic cleaning baths
  • Final rinse with distilled water will leave a residue-free surface
  • Phosphate and halogen free

Spécifications

Spécifications

Liquid
Off-White
Used on equipment and apparatus made of Glass, Plastic, Quartz, Porcelain, Ferrous Metals and Rubber
Videos
FDS
missing translation for 'documents'

missing translation for 'documents'