Learn More
MilliporeSigma™ Extran™ 300 Detergent
Powerful, liquid concentrate specially formulated to remove stubborn contamination and burnt-on residues
Supplier: MilliporeSigma™ EX09962
Description
Can be used on equipment and apparatus made of glass, plastic, quartz, porcelain, ferrous metals and rubber.
- Especially effective at removing organic materials such as blood and proteins
- Works well in ultrasonic cleaning baths
- Final rinse with distilled water will leave a residue-free surface
- Phosphate and halogen free
Specifications
| Liquid | |
| Poly bottle | |
| 1 L |
| Off-White | |
| Used on equipment and apparatus made of Glass, Plastic, Quartz, Porcelain, Ferrous Metals and Rubber |