Learn More
Eisco™ Polystyrene Slide Boxes
Durable, used for slide storage and organization
Supplier:  Eisco™ BI0114C
 Afficher plus de versions de ce produit
Numéro de catalogue. S95934C
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Durable, polystyrene box with slits to hold microscope slides
  • Compact design with heavy walls that will not warp, splinter or crack
  • Accommodates 75 x 25 mm slides
Spécifications
Spécifications
Slide Box
50
Heavy walls
50
Microscope Slide
Polystyrene
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS