missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EBF1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5150324

Numéro de catalogue. PA5150324

Il en reste null
Découvrez plus d'offres spéciales

Description

Description

EBF1 Polyclonal Antibody for IHC, Western Blot

Transcriptional activator which recognizes variations of the palindromic sequence 5'-ATTCCCNNGGGAATT-3'. Target protein is a transcription factor that may regulate several olfactory-specific genes.
Spécifications

Spécifications

EBF1
Polyclonal
Unconjugated
EBF1
COE1; Collier; Collier, Olf and EBF transcription factor 1; early B cell factor 1; early B-cell factor; early B-cell factor (olfactory neuronal transcription factor 1); early B-cell factor 1; Ebf; EBF transcription factor 1; EBF1; O/E-1; OE-1; OLF1; Olf-1; Olfactory neuronal transcription factor; olfactory neuronal transcription factor 1; transcription factor COE1
Rabbit
Protein A
RUO
116543, 13591, 1879
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry, Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q07802, Q63398, Q9UH73
EBF1
Recombinant Early B-Cell Factor 1 (EBF1) (Asp179-Ser451).
200 μL
Primary
Human, Mouse, Rat
Antibody
IgG
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions