Learn More
BUCHI Lyovapor™ Drying Chamber
Freeze drying flexibility for basic and advanced drying of vials, shelves, and flasks.
Supplier: BUCHI 11DCIN14200
Description
- Manifolds: adapters for traditional freeze drying beakers or round bottom flasks
- Shelves: for vials, plates, and irregular shaped items
- Heated Shelves: advanced control of drying process and applications
- Stoppering Chambers: seal vials without exposure to ambient moisture
Spécifications
| Benchtop Freeze Dryer | |
| Ambient to -105°C | |
| 12-position Manifold Top | |
| North America |
| 4 | |
| Acrylic | |
| Drying Chamber Only | |
| 220/240 V |