Learn More
Thermo Scientific Chemicals DL-Propranolol hydrochloride, 99%
CAS: 318-98-9 | C16H21NO2·HCl | 295.8 g/mol
Supplier: Thermo Scientific Chemicals 207321000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
| DL-Propranolol hydrochloride | |
| 163.0°C to 166.0°C | |
| Authentic | |
| Glass bottle | |
| C10H7OCH2CH(OH)CH2NHCH(CH3)2·HCl | |
| 100 g | |
| propranolol hydrochloride, propranolol hcl, inderal, avlocardyl, herzbase, inderalici, naprilin, pronovan, dociton, ikopal | |
| ZMRUPTIKESYGQW-UHFFFAOYSA-N | |
| 1-naphthalen-1-yloxy-3-(propan-2-ylamino)propan-2-ol;hydrochloride | |
| 62882 | |
| 295.8 | |
| Powder |
| 318-98-9 | |
| White to Yellow | |
| 99% | |
| C16H21NO2·HCl | |
| MFCD00012558 | |
| 15, 7953 | |
| Solubility in water: soluble. Other solubilities: soluble in alcohol, practically insoluble in ether, benzene and ethyl, acetate | |
| CC(C)NCC(COC1=CC=CC2=CC=CC=C21)O.Cl | |
| 295.8 | |
| CHEBI:8500 | |
| 99% |
Identifiants chimiques
| 318-98-9 | |
| 295.8 | |
| ZMRUPTIKESYGQW-UHFFFAOYSA-N | |
| 62882 | |
| 1-naphthalen-1-yloxy-3-(propan-2-ylamino)propan-2-ol;hydrochloride |
| C16H21NO2·HCl | |
| MFCD00012558 | |
| propranolol hydrochloride, propranolol hcl, inderal, avlocardyl, herzbase, inderalici, naprilin, pronovan, dociton, ikopal | |
| CHEBI:8500 | |
| CC(C)NCC(COC1=CC=CC2=CC=CC=C21)O.Cl |
Sécurité et manipulation
GHS H Statement
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 206-268-7
RUO – Research Use Only