missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Propranolol hydrochloride, 99%

Catalog No. p-3779504
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC207321000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC207321000 Supplier Thermo Scientific Chemicals Supplier No. 207321000
Only null left

CAS: 318-98-9 | C16H21NO2·HCl | 295.8 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 318-98-9
Molecular Formula C16H21NO2·HCl
Molecular Weight (g/mol) 295.8
MDL Number MFCD00012558
InChI Key ZMRUPTIKESYGQW-UHFFFAOYSA-N
Synonym propranolol hydrochloride, propranolol hcl, inderal, avlocardyl, herzbase, inderalici, naprilin, pronovan, dociton, ikopal
PubChem CID 62882
ChEBI CHEBI:8500
IUPAC Name 1-naphthalen-1-yloxy-3-(propan-2-ylamino)propan-2-ol;hydrochloride
SMILES CC(C)NCC(COC1=CC=CC2=CC=CC=C21)O.Cl

Specifications

Melting Point 163.0°C to 166.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C10H7OCH2CH(OH)CH2NHCH(CH3)2·HCl
Quantity 100 g
Merck Index 15, 7953
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, practically insoluble in ether, benzene and ethyl, acetate
Formula Weight 295.8
Percent Purity 99%
Physical Form Powder
Chemical Name or Material DL-Propranolol hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 206-268-7

RUO – Research Use Only