missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dithieno[3,2-c:2',3'-e]oxepine-4,6-dione 98.0+%, TCI America™
Supplier: TCI America D49725G
Spécifications
| Dithieno[3,2-c:2′,3′-e]oxepine-4,6-dione | |
| 263°C | |
| 5 g | |
| C1=CSC2=C1C(=O)OC(=O)C3=C2SC=C3 | |
| 236.259 | |
| 236.26 | |
| Crystalline Powder |
| 1043023-52-4 | |
| C10H4O3S2 | |
| UGFYLGZFYCTMFU-UHFFFAOYSA-N | |
| dithieno[2,3-a | |
| 89807128 | |
| ≥98.0% (GC,T) |
Identifiants chimiques
| 1043023-52-4 | |
| 236.259 | |
| 89807128 | |
| C1=CSC2=C1C(=O)OC(=O)C3=C2SC=C3 |
| C10H4O3S2 | |
| UGFYLGZFYCTMFU-UHFFFAOYSA-N | |
| dithieno[2,3-a |
Sécurité et manipulation
missing translation for 'tsca' : No