missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dithieno[3,2-c:2',3'-e]oxepine-4,6-dione 98.0+%, TCI America™
$527.52 - $1582.59
Chemical Identifiers
| CAS | 1043023-52-4 |
|---|---|
| Molecular Formula | C10H4O3S2 |
| Molecular Weight (g/mol) | 236.259 |
| InChI Key | UGFYLGZFYCTMFU-UHFFFAOYSA-N |
| PubChem CID | 89807128 |
| IUPAC Name | dithieno[2,3-a |
| SMILES | C1=CSC2=C1C(=O)OC(=O)C3=C2SC=C3 |
Chemical Identifiers
| 1043023-52-4 | |
| 236.259 | |
| 89807128 | |
| C1=CSC2=C1C(=O)OC(=O)C3=C2SC=C3 |
| C10H4O3S2 | |
| UGFYLGZFYCTMFU-UHFFFAOYSA-N | |
| dithieno[2,3-a |
Specifications
| 1043023-52-4 | |
| C10H4O3S2 | |
| UGFYLGZFYCTMFU-UHFFFAOYSA-N | |
| dithieno[2,3-a | |
| 89807128 | |
| ≥98.0% (GC,T) | |
| Dithieno[3,2-c:2′,3′-e]oxepine-4,6-dione |
| 263°C | |
| 1 g | |
| C1=CSC2=C1C(=O)OC(=O)C3=C2SC=C3 | |
| 236.259 | |
| 236.26 | |
| Crystalline Powder |
Safety and Handling
TSCA : No