missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-(+)-Cellotriose, 95%

Supplier:  Thermo Scientific Chemicals J67702MA

 Afficher plus de versions de ce produit

Numéro de catalogue. AAJ67702MA


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

A 3-glucose cellulodextrin derived from cellulose degradation. An energy source for cellulosic bacteria. Can be used to help identify, differentiate and characterize oligosaccharide metabolizing enzymes such as endoglucanases and --glucosidases. Also useful in saccharification and ethanol fermentation research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

D-(+)-Cellotriose
Plastic bottle
MFCD00069842
100353
cellotriose, d-+-cellotriose, polysaccharide, d-cellotriose, beta-d-glucopyranosyl-1->4-beta-d-glucopyranosyl-1->4-d-glucose, beta-d-glcp-1->4-beta-d-glcp-1->4-d-glc, beta-d-glucopyranosyl-1->4-beta-d-glucopyranosyl-1->4-d-glucopyranose, cellodextrin, d-glucose, o-d-glucopyranosyl, cellooligosaccharide
FYGDTMLNYKFZSV-ZWSAEMDYSA-N
(2S,3R,4S,5S,6R)-2-{[(2R,3S,4R,5R,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
5287993
504.44
Powder
33404-34-1
C18H32O16
10 mg
Hygroscopic
Soluble in water at 50mg/ml /n
OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O[C@H]3[C@H](O)[C@@H](O)C(O)O[C@@H]3CO)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O
504.44
CHEBI:3528
95%

Identifiants chimiques

33404-34-1
504.44
FYGDTMLNYKFZSV-ZWSAEMDYSA-N
5287993
(2S,3R,4S,5S,6R)-2-{[(2R,3S,4R,5R,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
C18H32O16
MFCD00069842
cellotriose, d-+-cellotriose, polysaccharide, d-cellotriose, beta-d-glucopyranosyl-1->4-beta-d-glucopyranosyl-1->4-d-glucose, beta-d-glcp-1->4-beta-d-glcp-1->4-d-glc, beta-d-glucopyranosyl-1->4-beta-d-glucopyranosyl-1->4-d-glucopyranose, cellodextrin, d-glucose, o-d-glucopyranosyl, cellooligosaccharide
CHEBI:3528
OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O[C@H]3[C@H](O)[C@@H](O)C(O)O[C@@H]3CO)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'tsca' : No

Recommended Storage : Store at -20°C; Store under Nitrogen

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only