missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanone, Technical, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor C15084LTGL

 Afficher plus de versions de ce produit

Numéro de catalogue. 18610110


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Spécifications

Spécifications

Cyclohexanone
1
155.6°C
Amber Glass Bottle
Technical
108-94-1
Clear, Colorless
4 L
98.14

Identifiants chimiques

108-94-1
98.14
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.