missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanone, Technical, Spectrum™ Chemical

Catalog No. p-7165384
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
55 gal.
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610111 500 mL Amber Glass Bottle
18610108 1 L Amber Glass Bottle
18610110 4 L Amber Glass Bottle
18610109 20 L Amber Glass Bottle
18610112 55 gal. Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18610111 Supplier Spectrum Chemical Mfg Cor Supplier No. C1508500MLGL
Only null left

Chemical Identifiers

CAS 108-94-1
Molecular Weight (g/mol) 98.14

Specifications

CAS Max % 1
Appearance Clear, Colorless
Boiling Point 155.6°C
Quantity 500 mL
Packaging Amber Glass Bottle
Grade Technical
Chemical Name or Material Cyclohexanone

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.