missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Crystal Violet, certified

CAS: 548-62-9 | C25H30ClN3 | 407.986 g/mol

Spécifications

Chemical Name or Material Crystal Violet
CAS 548-62-9 , 90-94-8
Absorption Ratio 0.98 to 1.2 (P-15/P+15)
Dye Content 88% min.
Molecular Formula C25H30ClN3
voir les spécifications

Produits 3
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
AC405830250
Afficher les documents Détails de la promotion
Thermo Scientific Chemicals
405830250
25 g N/A
Il en reste null
 
AC405831000
Afficher les documents Détails de la promotion
Thermo Scientific Chemicals
405831000
100 g N/A
Il en reste null
 
AC405835000
Afficher les documents Détails de la promotion
Thermo Scientific Chemicals
405835000
500 g N/A
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Spécifications

Spécifications

Crystal Violet
0.98 to 1.2 (P-15/P+15)
C25H30ClN3
13, 756
Basic Violet 3, C.I. 42555, Gentian Violet
Authentic
[4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
11057
407.99
Green
548-62-9 , 90-94-8
88% min.
MFCD00011750
15, 4430
ZXJXZNDDNMQXFV-UHFFFAOYSA-M
CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
407.986
CHEBI:41688
Certified
Crystalline Powder
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye damage.
May cause cancer.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Do not eat, drink or smoke when using this product.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

FDS
Documents

Documents

RUO – Research Use Only