Learn More
Thermo Scientific Chemicals Crystal Violet, certified
CAS: 548-62-9 | C25H30ClN3 | 407.986 g/mol
Supplier: Thermo Scientific Chemicals 405831000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
| 548-62-9 , 90-94-8 | |
| 88% min. | |
| MFCD00011750 | |
| 15, 4430 | |
| ZXJXZNDDNMQXFV-UHFFFAOYSA-M | |
| CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-] | |
| 407.986 | |
| CHEBI:41688 | |
| Certified | |
| Crystalline Powder |
| 0.98 to 1.2 (P-15/P+15) | |
| C25H30ClN3 | |
| 13, 756 | |
| Basic Violet 3, C.I. 42555, Gentian Violet | |
| Authentic | |
| [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride | |
| 11057 | |
| 407.99 | |
| Green | |
| Crystal Violet |
Specifications
| Crystal Violet | |
| 548-62-9 , 90-94-8 | |
| 88% min. | |
| MFCD00011750 | |
| 15, 4430 | |
| Solubility in water: 16g/L (25°C). Other solubilities: soluble in chloroform, 1 g/10 mL alcohol, 1 g/15 mL glycerin, practically insoluble in ether | |
| CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-] | |
| [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride | |
| 11057 | |
| 407.99 | |
| Glass bottle | |
| 14.1 | |
| 100 g |
| 215.0°C | |
| 0.98 to 1.2 (P-15/P+15) | |
| C25H30ClN3 | |
| 13, 756 | |
| Basic Violet 3, C.I. 42555, Gentian Violet | |
| ZXJXZNDDNMQXFV-UHFFFAOYSA-M | |
| Authentic | |
| 407.986 | |
| CHEBI:41688 | |
| Certified | |
| >215°C | |
| Green | |
| Crystalline Powder |
Safety and Handling
GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye damage.
May cause cancer.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Do not eat, drink or smoke when using this product.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only