missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Crystal Violet, certified
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
| Chemical Name or Material | Crystal Violet |
| CAS | 548-62-9, 90-94-8 |
| Absorption Ratio | 0.98 to 1.2 (P-15/P+15) |
| Dye Content | 88% min. |
| Infrared Spectrum | Authentic |
| Molecular Formula | C25H30ClN3 |
| MDL Number | MFCD00011750 |
| Beilstein | 13, 756 |
| Merck Index | 15, 4430 |
| Color | Green |
| Show More |
RUO – Research Use Only