Learn More
Corning™ Reusable Plastic Erlenmeyer Flasks with Red Graduations
Translucent polymethylpentene; with polypropylene screw cap
Supplier:  Corning™ 4990P500
 Afficher plus de versions de ce produit
Numéro de catalogue. 07-202-118
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
Translucent polymethylpentene flasks are manufactured with red graduations for reference. The flasks have a screw neck and are supplied with a PP cap.
Spécifications
Spécifications
Polypropylene
Polypropylene
Narrow Mouth
1/Pk.
500 mL
100 mL
6/Cs.
Screw Cap
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS