missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper Sulfate, For Morpholine, 2g/L, LabChem™
Supplier: LabChem LC134101
Spécifications
| Copper Sulfate | |
| For Morpholine | |
| 99.8,0.2 | |
| CuSO4·5H2O | |
| 2g/L | |
| copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas | |
| JZCCFEFSEZPSOG-UHFFFAOYSA-L | |
| copper(2+) pentahydrate sulfate | |
| 24463 | |
| 249.68 | |
| Poly Bottle | |
| Sulfur compounds | |
| Liquid |
| Blue | |
| 7758-99-8 , 7732-18-5 | |
| CuH10O9S | |
| MFCD00149681 | |
| 1g/mL | |
| Soluble in water | |
| O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O | |
| 249.68 | |
| CHEBI:31440 | |
| Passes Test | |
| 1g/mL | |
| 500 mL |
Identifiants chimiques
| 7758-99-8 | |
| 249.68 | |
| JZCCFEFSEZPSOG-UHFFFAOYSA-L | |
| 24463 | |
| copper(2+) pentahydrate sulfate |
| CuH10O9S | |
| MFCD00149681 | |
| copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas | |
| CHEBI:31440 | |
| O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O |
Sécurité et manipulation
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature