missing translation for 'onlineSavingsMsg'
Learn More

CLEC11A Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Goat Polyclonal Antibody

$337.00 - $432.00

Spécifications

Antigen CLEC11A
Concentration 0.1-1.0 mg/mL
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Biotin
voir les spécifications

Produits 3
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
500P99GBT25 DFS Item
Afficher les documents
PeproTech
500P99GBT25UG
25 μg
chaque for $337.00
Il en reste null
 
500P99GBT50 DFS Item
Afficher les documents
PeproTech
500P99GBT50UG
50 μg
chaque for $432.00
Il en reste null
 
50-271-8426
Afficher les documents
PeproTech
500P99GBT500UG
500 μg N/A
N/A
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Description

Description

AA Sequence of recombinant protein: MGHGARGAER EWEGGWGGAQ EEEREREALM LKHLQEALGL PAGRGDENPA GTVEGKEDWE MEEDQGEEEE EEATPTPSSG PSPSPTPEDI VTYILGRLAG LDAGLHQLHV RLHALDTRVV ELTQGLRQLR NAAGDTRDAV QALQEAQGRA EREHGRLEGC LKGLRLGHKC FLLSRDFEAQ PSASPHPLSP DQPNGGTLEN CVAQASDDGS WWDHDCQRRL YYVCEFPF. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human SCGF-beta. Anti-Human SCGF-beta-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hSCGF-beta by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human SCGF-beta (500-P99G) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hSCGF-beta. Western Blot: To detect hSCGF-beta by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hSCGF-beta is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P99GBT-1MG will be provided as 2 x 500 μg

This gene encodes a member of the C-type lectin superfamily. The encoded protein is a secreted sulfated glycoprotein and functions as a growth factor for primitive hematopoietic progenitor cells. An alternative splice variant has been described but its biological nature has not been determined.
Spécifications

Spécifications

CLEC11A
ELISA, Western Blot
Biotin
Goat
Human
Q9Y240
6320
E.coli-derived, 25.0 kDa Recombinant Human SCGF-beta.
Antigen affinity chromatography
CLEC11A
0.1-1.0 mg/mL
Polyclonal
Lyophilized
RUO
PBS with no preservative
AW457320; b SCGF; beta SCGF; betaSCGF; bSCGF; Clec11a; CLECSF3; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 3; C-type lectin domain family 11 member A; C-type lectin domain family 11, member A; C-type lectin superfamily member 3; hCG_22930; LSLCL; lymphocyte secreted C-type lectin; lymphocyte secreted long form of C-type lectin; Osteolectin; P47; SCGF; SCGF b; SCGF beta; SCGF β; SCGFb; SCGFbeta; SCGFβ; stem cell growth factor; stem cell growth factor; lymphocyte secreted C-type lectin; β SCGF; βSCGF
CLEC11A
Primary
-20°C
Videos
FDS
missing translation for 'documents'

missing translation for 'documents'